Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 12 | 2834 | ||
GenBank mRNA | AK032204.1 | - | 12 | 2492 | ||
AK034077.1 | - | 12 | 2452 | |||
AK034240.1 | - | 12 | 2487 | |||
AK038630.1 | - | 1 | 643 | |||
AK045621.1 | - | 1 | 454 | |||
AK047702.1 | - | 1 | 234 | |||
AY164459.1 | - | 12 | 2834 | |||
BC075646.1 | - | 12 | 2464 | 30620475 |