Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178706.4 | NP_848821.2 | 6 | 1317 | ||
GenBank mRNA | AK032377.1 | - | 5 | 1222 | ||
AK045719.1 | - | 7 | 947 | |||
AK047813.1 | - | 6 | 1242 |