Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252503.2 | NP_001239432.1 | 6 | 2575 | ||
NM_001290545.1 | NP_001277474.1 | 6 | 2575 | |||
NM_181411.4 | NP_852076.1 | 6 | 2575 | |||
GenBank mRNA | AK029681.1 | - | 6 | 2474 | ||
AK030572.1 | - | 6 | 2519 | |||
AK047225.1 | - | 5 | 1639 | |||
AK136815.1 | - | 5 | 1827 | |||
AK150458.1 | - | 4 | 366 | |||
AK152308.1 | - | 4 | 366 | |||
AK152487.1 | - | 4 | 366 | |||
AK161532.1 | - | 4 | 379 | |||
BC004630.1 | - | 5 | 864 | |||
BC039995.1 | - | 1 | 33 | |||
BC052036.1 | - | 6 | 2503 | 5702534 |