Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181593.2 | NP_853624.1 | 1 | 447 | ||
GenBank mRNA | AK033605.1 | - | 1 | 117 | ||
AK154888.1 | - | 1 | 479 | |||
BC013227.1 | - | 1 | 333 | |||
BC053450.1 | - | 1 | 447 | 6508198 |