Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rerg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164212.1 | NP_001157684.1 | 5 | 2168 | ||
NM_001164214.1 | NP_001157686.1 | 4 | 2111 | |||
NM_181988.2 | NP_871788.1 | 5 | 1988 | |||
GenBank mRNA | AK133734.1 | - | 5 | 2150 | ||
BC026463.1 | - | 5 | 2006 | 4483442 |