Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182783.2 | NP_877584.2 | 2 | 789 | ||
GenBank mRNA | AK134621.1 | - | 2 | 619 | ||
AK143928.1 | - | 2 | 759 | |||
BC030183.1 | - | 2 | 756 | |||
BX517339.1 | - | 2 | 372 |