Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183017.2 | NP_898838.2 | 10 | 1491 | ||
GenBank mRNA | AK084879.1 | - | 10 | 1458 | ||
AK134345.1 | - | 10 | 1458 | |||
AK143496.1 | - | 9 | 1324 | |||
AK150116.1 | - | 10 | 1457 | |||
AM690756.1 | - | 10 | 1444 | |||
BC055368.1 | - | 10 | 1456 | 30055801 |