Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183019.2 | NP_898840.2 | 3 | 567 | ||
GenBank mRNA | AK031310.1 | - | 3 | 550 | ||
AK034013.1 | - | 3 | 531 | |||
AK134629.1 | - | 530 | ||||
AK138598.1 | - | 3 | 522 | |||
AK163424.1 | - | 3 | 549 | |||
AK169449.1 | - | 3 | 550 | |||
AK169957.1 | - | 3 | 587 | |||
BB609907.1 | - | 3 | 569 | |||
BC055804.1 | - | 3 | 529 | |||
BC094608.1 | - | 4087 | ||||
BC139371.1 | - | 3 | 550 | 8862393 | ||
BC139397.1 | - | 3 | 550 | 8862419 |