Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194340.2 | NP_919321.2 | 6 | 5143 | ||
GenBank mRNA | AY372183.1 | - | 1 | 364 | ||
BC094374.1 | - | 1 | 690 | 30531815 |