Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194340.2 | NP_919321.2 | 11 | 6299 | ||
GenBank mRNA | AK049285.1 | - | 4 | 632 | ||
AY372183.1 | - | 6 | 1520 | |||
BC059908.1 | - | 4 | 731 | |||
BC094374.1 | - | 6 | 1846 | 30531815 |