Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198248.1 | NP_937891.1 | 2 | 633 | ||
GenBank mRNA | AK036584.1 | - | 2 | 739 | ||
AK220256.1 | - | 3 | 518 | |||
BC059177.1 | - | 2 | 633 | 6410689 |