Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198302.2 | NP_938044.1 | 3 | 348 | ||
GenBank mRNA | AK039297.1 | - | 3 | 290 | ||
BC050779.1 | - | 3 | 395 | 6771154 |