Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198305.2 | NP_938047.1 | 5 | 885 | ||
GenBank mRNA | AK049214.1 | - | 5 | 762 | ||
AK154523.1 | - | 5 | 823 | |||
AK170825.1 | - | 5 | 823 | |||
AY423764.1 | - | 5 | 863 | |||
BC139309.1 | - | 5 | 826 | 8862331 |