Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198422.2 | NP_940814.1 | 5 | 932 | ||
GenBank mRNA | AK149213.1 | - | 5 | 911 | ||
AK172689.1 | - | 5 | 888 | |||
AY424292.1 | - | 5 | 932 | |||
BC120853.1 | - | 5 | 917 | 40129776 | ||
BC120855.1 | - | 5 | 918 | 40129778 |