Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199015.4 | NP_950180.3 | 6, 7 | 614 | ||
GenBank mRNA | AK156583.1 | - | 6, 7 | 595 | ||
BC030186.1 | - | 6, 7 | 615 | 5067268 | ||
BC110663.1 | - | 5, 6 | 1698 | |||
BC145673.1 | - | 6, 7 | 534 | 40130976 | ||
EU155107.1 | - | 3, 4 | 225 |