Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199016.2 | NP_950181.2 | 3 | 1037 | ||
GenBank mRNA | AK090098.1 | - | 3 | 1038 | ||
AK157622.1 | - | 3 | 933 | |||
AK173048.1 | - | 4 | 997 | |||
BC027749.1 | - | 3 | 991 | 5065563 | ||
CB723093.1 | - | 2 | 486 |