Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010620.1 | NP_034750.1 | 4 | 357 | ||
GenBank mRNA | AJ560623.1 | - | 4 | 374 | ||
AK050821.1 | - | 4 | 374 | |||
BC060710.1 | - | 4 | 357 | 6409082 |