Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012026.2 | NP_036156.2 | 5 | 691 | ||
GenBank mRNA | AK173325.1 | - | 5 | 714 | ||
BC022978.1 | - | 5 | 619 | |||
BC057883.1 | - | 5 | 658 | |||
BC075734.1 | - | 5 | 679 | |||
BC098331.1 | - | 5 | 680 | |||
BC138030.1 | - | 5 | 615 | 8861049 | ||
U73199.1 | - | 5 | 691 |