Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022317.3 | NP_071712.3 | 2 | 198 | ||
GenBank mRNA | AF305211.1 | - | 2 | 131 | ||
AK052784.1 | - | 2 | 199 | |||
AK085680.1 | - | 3 | 627 | |||
AK145077.1 | - | 2 | 199 | |||
AK164740.1 | - | 3 | 627 | |||
AK164752.1 | - | 2 | 199 | |||
BC010472.1 | - | 2 | 186 | 3586005 | ||
BC013783.1 | - | 2 | 189 | 3600786 | ||
BC037061.1 | - | 2 | 201 | |||
BY736278.1 | - | 2 | 199 |