Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023377.4 | NP_075866.2 | 3 | 221 | ||
GenBank mRNA | AF480302.1 | - | 3 | 197 | ||
AK009980.1 | - | 3 | 227 | |||
AK077328.1 | - | 3 | 239 | |||
AK080060.1 | - | 3 | 239 | |||
AK085852.1 | - | 3 | 234 | |||
AK142503.1 | - | 3 | 239 | |||
AK158178.1 | - | 3 | 223 | |||
AY007808.1 | - | 3 | 216 | |||
BC031733.1 | - | 3 | 225 | |||
BC058226.1 | - | 3 | 221 | 6308417 |