Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025438.2 | NP_001020609.1 | 19 | 2176 | ||
NM_001025439.2 | NP_001020610.1 | 20 | 2189 | |||
NM_001293663.1 | NP_001280592.1 | 20 | 2189 | |||
NM_001293664.1 | NP_001280593.1 | 19 | 2129 | |||
NM_001293665.1 | NP_001280594.1 | 20 | 2169 | |||
NM_023813.4 | NP_076302.1 | 18 | 2087 | |||
GenBank mRNA | AK012702.1 | - | 18 | 1708 | ||
AK039076.1 | - | 20 | 1798 | |||
AK044264.1 | - | 12 | 1115 | |||
AK132185.1 | - | 5 | 606 | |||
AK148840.1 | - | 19 | 1836 | |||
AK220516.1 | - | 20 | 2189 | |||
BC042895.1 | - | 19 | 2717 | |||
BC052894.1 | - | 19 | 1787 | 30055337 | ||
BC057103.1 | - | 20 | 2085 |