Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024236.2 | NP_077198.1 | 7 | 840 | ||
GenBank mRNA | AK011351.1 | - | 7 | 769 | ||
AK078123.1 | - | 7 | 793 | |||
AK159107.1 | - | 7 | 784 | |||
AK161660.1 | - | 7 | 733 | |||
AK166694.1 | - | 7 | 794 | |||
BC002107.1 | - | 7 | 780 | 3487104 |