Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276442.1 | NP_001263371.1 | 3, 4 | 266 | ||
NM_025371.3 | NP_079647.1 | 3, 4 | 270 | |||
GenBank mRNA | AK002423.1 | - | 3, 4 | 256 | ||
AK003703.1 | - | 3, 4 | 271 | |||
BC005631.1 | - | 3, 4 | 274 | 3964138 | ||
CV560337.1 | - | 3, 4 | 191 |