Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 2 | 279 | ||
XM_017026240.1 | XP_016881729.1 | 1 | 164 | |||
GenBank mRNA | AK129878.1 | - | 2 | 177 | ||
AK300730.1 | - | 2 | 177 | |||
AK313281.1 | - | 2 | 177 | |||
BC007462.1 | - | 2 | 177 | 3934232 | ||
BT006793.1 | - | 1 | 104 | |||
DA896166.1 | - | 2 | 277 | |||
DC385783.1 | - | 2 | 280 | |||
M14780.1 | - | 2 | 182 |