Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252374.1 | NP_001239303.1 | 3 | 132 | ||
NM_026004.3 | NP_080280.3 | 2 | 289 | |||
GenBank mRNA | AK011525.1 | - | 2 | 240 | ||
AK011894.1 | - | 2 | 187 | |||
AK160139.1 | - | 3 | 132 | |||
AK208033.1 | - | 2 | 52 | |||
BC038029.1 | - | 3 | 125 | 4018956 | ||
BY080680.1 | - | 2 | 291 | |||
CB571852.1 | - | 3 | 137 |