Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026240.2 | NP_080516.2 | 5 | 572 | ||
GenBank mRNA | AK018551.1 | - | 6 | 822 | ||
AK018691.1 | - | 5 | 558 | |||
AK038289.1 | - | 4 | 613 | |||
AK161329.1 | - | 5 | 505 | |||
AK164050.1 | - | 5 | 557 | |||
BC057978.1 | - | 5 | 547 | 5044170 |