Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026240.2 | NP_080516.2 | 6, 7 | 732 | ||
GenBank mRNA | AK018551.1 | - | 7, 8 | 982 | ||
AK018691.1 | - | 6, 7 | 718 | |||
AK038289.1 | - | 5, 6 | 773 | |||
AK161329.1 | - | 6, 7 | 665 | |||
AK164050.1 | - | 6, 7 | 717 | |||
BC057978.1 | - | 6, 7 | 707 | 5044170 |