Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026695.3 | NP_080971.2 | 5 | 641 | ||
GenBank mRNA | AK002407.1 | - | 5 | 583 | ||
AK013284.1 | - | 5 | 605 | |||
AK150293.1 | - | 5 | 584 | |||
BC049237.1 | - | 5 | 570 | 4241552 | ||
BC069877.1 | - | 5 | 600 | 5370580 |