Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026887.3 | NP_081163.2 | 5 | 1057 | ||
GenBank mRNA | AK048085.1 | - | 1 | 480 | ||
AK053941.1 | - | 5 | 930 | |||
BC046964.1 | - | 5 | 914 | 6485320 |