Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 12 | 1723 | ||
GenBank mRNA | AF306557.1 | - | 12 | 1727 | ||
AK036505.1 | - | 13 | 1823 | |||
AK047057.1 | - | 12 | 1716 | |||
AK142165.1 | - | 12 | 1688 | |||
AK154172.1 | - | 12 | 1705 | |||
AK166022.1 | - | 12 | 1706 | |||
AK166340.1 | - | 12 | 1721 | |||
AK170290.1 | - | 12 | 1720 | |||
BC031409.1 | - | 12 | 1666 | 4952710 |