Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027828.2 | NP_082104.2 | 1 | 589 | ||
GenBank mRNA | AK018545.1 | - | 1 | 564 | ||
AK139763.1 | - | 1 | 697 | |||
AK160079.1 | - | 1 | 593 | |||
BC019466.1 | - | 1 | 589 | 4458596 | ||
BC085288.1 | - | 1 | 574 | 5354365 |