Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028133.2 | NP_082409.2 | 5 | 1493 | ||
GenBank mRNA | AF421882.1 | - | 5 | 1395 | ||
AJ310548.1 | - | 5 | 1474 | |||
AK011502.1 | - | 4 | 797 | |||
AK044787.1 | - | 5 | 1495 | |||
AK165972.1 | - | 5 | 1424 | |||
AK170732.1 | - | 5 | 1424 | |||
BC044926.1 | - | 5 | 1403 | 5373240 | ||
BC058278.1 | - | 5 | 1413 | 6528802 |