Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005969.3 | NP_005960.1 | 4 | 275 | ||
GenBank mRNA | AB500094.1 | - | 2 | 134 | ||
AK299515.1 | - | 4 | 198 | |||
AK312599.1 | - | 4 | 283 | |||
AK316242.1 | - | 4 | 192 | |||
AK316548.1 | - | 5 | 332 | |||
BC022090.2 | - | 4 | 185 | 3029455 | ||
U77456.1 | - | 4 | 283 |