Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252316.1 | NP_001239245.1 | 8 | 1059 | ||
NM_028230.4 | NP_082506.1 | 8 | 1068 | |||
GenBank mRNA | AK012355.1 | - | 8 | 1013 | ||
AK037339.1 | - | 8 | 1022 | |||
AK051207.1 | - | 8 | 1059 | |||
AK162421.1 | - | 8 | 1069 | |||
AK169192.1 | - | 8 | 1002 | |||
AK194812.1 | - | 1 | 68 | |||
AK207979.1 | - | 1 | 68 | |||
BC004825.1 | - | 8 | 1027 | 2615850 | ||
BC051396.1 | - | 8 | 990 | 5132676 |