Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028266.5 | NP_082542.3 | 6, 7 | 928 | ||
GenBank mRNA | AK040971.1 | - | 6, 7 | 881 | ||
AK051511.1 | - | 6, 7 | 884 | |||
AK142403.1 | - | 6, 7 | 860 | |||
BC027766.1 | - | 6, 7 | 815 | 5346676 |