Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289648.1 | NP_001276577.1 | 7 | 646 | ||
NM_001289649.1 | NP_001276578.1 | 7 | 625 | |||
NM_001289650.1 | NP_001276579.1 | 7 | 660 | |||
NM_029019.4 | NP_083295.1 | 7 | 715 | |||
GenBank mRNA | AF480303.1 | - | 4 | 362 | ||
AK014759.1 | - | 7 | 626 | |||
AK016984.1 | - | 7 | 626 | |||
BC061022.1 | - | 5 | 398 | 6774873 | ||
BC087893.1 | - | 7 | 637 | 6774832 | ||
JF440953.1 | - | 5 | 434 | |||
JF440954.1 | - | 5 | 434 |