Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167914.1 | NP_001161386.1 | 6, 7 | 562 | ||
NM_029705.3 | NP_083981.2 | 6, 7 | 562 | |||
GenBank mRNA | AF537188.1 | - | 6, 7 | 476 | ||
AK008675.1 | - | 6, 7 | 521 | |||
AK030842.1 | - | 6, 7 | 502 | |||
BC049743.1 | - | 6, 7 | 562 | |||
BC087880.1 | - | 6, 7 | 557 | 6771498 |