Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005980.2 | NP_005971.1 | 2 | 306 | ||
GenBank mRNA | AF539739.1 | - | 2 | 1076 | ||
AY423724.1 | - | 2 | 240 | |||
BC006819.1 | - | 2 | 299 | 3448490 | ||
BI259919.1 | - | 2 | 318 | |||
BT007289.1 | - | 2 | 212 | |||
X65614.1 | - | 2 | 233 |