Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030201.3 | NP_084477.1 | 2 | 189 | ||
GenBank mRNA | AK021006.1 | - | 2 | 161 | ||
AK030553.1 | - | 2 | 162 | |||
AK034643.1 | - | 2 | 169 | |||
AK049011.1 | - | 2 | 157 | |||
AK167839.1 | - | 2 | 132 | |||
BC039701.1 | - | 1 | 130 | |||
BC085181.1 | - | 2 | 149 | 30646642 | ||
BC094508.1 | - | 2 | 132 | |||
BY133105.1 | - | 2 | 190 |