Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030206.4 | NP_084482.1 | 4 | 1439 | ||
GenBank mRNA | AK019410.1 | - | 4 | 1440 | ||
AK087408.1 | - | 4 | 1425 | |||
AK163073.1 | - | 4 | 1190 | |||
AK172475.1 | - | 4 | 1186 | |||
BC055040.1 | - | 6 | 1348 | 6491041 |