Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030693.2 | NP_109618.1 | 3 | 484 | ||
GenBank mRNA | AF310624.1 | - | 3 | 505 | ||
AF312938.1 | - | 3 | 460 | |||
AF375475.1 | - | 3 | 489 | |||
AF375476.1 | - | 3 | 484 | |||
AK131939.1 | - | 3 | 505 | |||
AK146479.1 | - | 3 | 506 | |||
AK159624.1 | - | 3 | 508 | |||
BC010195.1 | - | 2 | 317 | 3157499 | ||
BC092068.1 | - | 3 | 489 | 4210931 |