Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033074.3 | NP_149065.2 | 3 | 320 | ||
GenBank mRNA | AK010256.1 | - | 3 | 287 | ||
AK011146.2 | - | 3 | 321 | |||
AK031064.1 | - | 3 | 284 | |||
AK152945.1 | - | 3 | 281 | |||
AK153314.1 | - | 3 | 292 | |||
AK166693.1 | - | 3 | 287 | |||
AK167597.1 | - | 3 | 329 | |||
AK168969.1 | - | 3 | 280 | |||
AK197272.1 | - | 2 | 126 | |||
AK206659.1 | - | 2 | 126 | |||
AK210237.1 | - | 2 | 126 | |||
BC055371.1 | - | 3 | 306 | 30060749 | ||
BC060176.1 | - | 263 |