Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033074.3 | NP_149065.2 | 9, 10 | 1111 | ||
GenBank mRNA | AK010256.1 | - | 9, 10 | 1078 | ||
AK011146.2 | - | 9, 10 | 1112 | |||
AK031064.1 | - | 9, 10 | 1075 | |||
AK152945.1 | - | 9, 10 | 1072 | |||
AK153314.1 | - | 9, 10 | 1083 | |||
AK166693.1 | - | 9, 10 | 1078 | |||
AK167597.1 | - | 9, 10 | 1120 | |||
AK168969.1 | - | 9, 10 | 1071 | |||
AK184579.1 | - | 1, 2 | 110 | |||
BC004621.1 | - | 2, 3 | 198 | |||
BC055371.1 | - | 9, 10 | 1097 | 30060749 | ||
BC060176.1 | - | 1127 |