Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033521.3 | NP_277056.1 | 7 | 1079 | ||
GenBank mRNA | AK077269.1 | - | 7 | 942 | ||
AK077933.1 | - | 7 | 1079 | |||
AK079581.1 | - | 7 | 942 | |||
AK082807.1 | - | 7 | 958 | |||
AK146868.1 | - | 6 | 845 | |||
AK166014.1 | - | 7 | 981 | |||
AK196114.1 | - | 1 | 89 | |||
AK198753.1 | - | 1 | 95 | |||
AK200493.1 | - | 1 | 89 | |||
BC019120.1 | - | 7 | 949 | 4507607 | ||
EU561010.1 | - | 7 | 1079 |