Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033523.4 | NP_277058.1 | 6 | 1411 | ||
GenBank mRNA | AB063496.1 | - | 6 | 922 | ||
AK036430.1 | - | 6 | 1412 | |||
AK157817.1 | - | 6 | 1414 | |||
BC040462.1 | - | 1 | 126 | |||
BC051247.1 | - | 1 | 355 | |||
BC066013.1 | - | 6 | 1481 | 5706237 |