Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033524.3 | NP_277059.1 | 7 | 1679 | ||
GenBank mRNA | AB063495.1 | - | 7 | 1025 | ||
AK155065.1 | - | 7 | 1642 | |||
AK155070.1 | - | 7 | 1682 | |||
BC041685.1 | - | 6 | 993 | |||
BC079606.1 | - | 7 | 1300 | 30355455 |