Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033612.2 | NP_291090.2 | 8 | 909 | ||
GenBank mRNA | AK007392.1 | - | 8 | 907 | ||
BC011218.1 | - | 8 | 883 | 4208442 | ||
M27347.1 | - | 7 | 791 |