Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197321.1 | NP_001184250.1 | 16 | 1907 | ||
NM_001197322.1 | NP_001184251.1 | 13 | 1514 | |||
NM_053202.2 | NP_444432.1 | 17 | 2059 | |||
GenBank mRNA | AB221601.1 | - | 14 | 1750 | ||
AB221602.1 | - | 14 | 1752 | |||
AB594192.1 | - | 18 | 2464 | |||
AF339103.1 | - | 14 | 1748 | |||
AF339105.1 | - | 1528 | ||||
AK033368.1 | - | 11 | 1224 | |||
AK077062.1 | - | 13 | 1409 | |||
AK087748.1 | - | 1 | 80 | |||
AK160315.1 | - | 5 | 511 | |||
BC064764.1 | - | 16 | 1893 | 30430449 |