Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054049.2 | NP_473390.1 | 2 | 1096 | ||
GenBank mRNA | AF370121.1 | - | 2 | 1096 | ||
AK052147.1 | - | 2 | 1111 | |||
AK161267.1 | - | 2 | 1111 | |||
AY038074.1 | - | 1 | 629 | |||
BC013504.1 | - | 2 | 903 | 4224339 |