Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054051.1 | NP_473392.1 | 10 | 3122 | ||
GenBank mRNA | AK089414.1 | - | 10 | 3114 | ||
AK142164.1 | - | 1 | 1686 | |||
AK146559.1 | - | 10 | 3175 | |||
AK170392.1 | - | 11 | 2605 | |||
AK171497.1 | - | 10 | 3120 | |||
BC047282.1 | - | 10 | 3122 | 6491155 |